3-Ethoxy-2-fluorbenzonitril
Glass bottle for solids, 5 g
Katalognr. COBBOT-0032-5G
Marketsource
Anbieter: COMBI-BLOCKS
3-Ethoxy-2-fluorbenzonitril
Katalognr. COBBOT-0032-5G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 1033202-20-8
Direkt zu
Product Details & Documents
- CAS Nummer:1033202-20-8
- Formel:C₉H₈FNO
- MDL-Nummer:MFCD10699689
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



