4-Bromchinolin
Glasflasche für Feststoffe, 5 g
Catalog # COBBOT-8087-5G
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:3964-04-3
- Formula:C₉H₆BrN
- MDL Number:MFCD07644514
- Molecular Weight:208.06
- Storage Temperature:Kühlschrank
- UN:3261
- ADR:8,II
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



