3-Chlor-5-nitrobenzonitril
Glass bottle for solids, 1 g
Katalognr. COBBCA-4299-1G
Marketsource
Anbieter: COMBI-BLOCKS
3-Chlor-5-nitrobenzonitril
Katalognr. COBBCA-4299-1G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 34662-30-1
Direkt zu
Product Details & Documents
- CAS Nummer:34662-30-1
- Formel:C₇H₃ClN₂O₂
- MDL-Nummer:MFCD00463805
- Molekulargewicht:182.57 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



