2-Fluor-4-nitroanisol
Glass bottle for solids, 100 g
Catalog # COBBAN-1093-100G
Marketsource
Supplier: COMBI-BLOCKS
2-Fluor-4-nitroanisol
Catalog # COBBAN-1093-100G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 455-93-6
Jump to:
Product Details & Documents
- CAS Number:455-93-6
- Formula:C₇H₆FNO₃
- MDL Number:MFCD00061095
- Molecular Weight:171.13 g/mol
- Storage Temperature:Raumtemperatur
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



