2-Fluor-4-nitroanisol
Glass bottle for solids, 25 g
Katalognr. COBBAN-1093-25G
Marketsource
Anbieter: COMBI-BLOCKS
2-Fluor-4-nitroanisol
Katalognr. COBBAN-1093-25G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 455-93-6
Direkt zu
Product Details & Documents
- CAS Nummer:455-93-6
- Formel:C₇H₆FNO₃
- MDL-Nummer:MFCD00061095
- Molekulargewicht:171.13 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



