1-Boc-2-indolboronsäure
Glass bottle for solids, 25 g
Katalognr. COBBBB-2009-25G
Marketsource
Anbieter: COMBI-BLOCKS
1-Boc-2-indolboronsäure
Katalognr. COBBBB-2009-25G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 213318-44-6
Direkt zu
Product Details & Documents
Synonyme: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
- CAS Nummer:213318-44-6
- Formel:C₁₃H₁₆BNO₄
- MDL-Nummer:MFCD02093045
- Molekulargewicht:261.08 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g



