1-Boc-2-indolboronsäure
Glass bottle for solids, 5 g
Catalog # COBBBB-2009-5G
Marketsource
Supplier: COMBI-BLOCKS
1-Boc-2-indolboronsäure
Catalog # COBBBB-2009-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 213318-44-6
Jump to:
Product Details & Documents
Synonyms: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
- CAS Number:213318-44-6
- Formula:C₁₃H₁₆BNO₄
- MDL Number:MFCD02093045
- Molecular Weight:261.08 g/mol
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



