3-Fluor-4-nitrophenol
Glasflasche für Feststoffe, 25 g
COBBAN-1020-25G
Marketsource
- :394-41-2
- :C₆H₄FNO₃
- :MFCD00041251
- :157.1 g/mol
- :90…92 °C
- :Raumtemperatur
- :206-895-6
- Packung:Glasflasche für Feststoffe
- VE:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.