3-Fluor-4-nitrophenol
Glasflasche für Feststoffe, 5 g
Katalognr. COBBAN-1020-5G
Marketsource
Anbieter: COMBI-BLOCKS
3-Fluor-4-nitrophenol
Katalognr. COBBAN-1020-5G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 394-41-2
Direkt zu
Product Details & Documents
- CAS Nummer:394-41-2
- Formel:C₆H₄FNO₃
- MDL-Nummer:MFCD00041251
- Molekulargewicht:157.1 g/mol
- Schmelzender Platingehalt:90…92 °C
- Lagertemperatur:Raumtemperatur
- EINECS:206-895-6
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
- Cat. No.:COBBAN-1020-5G
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



