2-Hydroxy-3-nitropyridin
Glass bottle for solids, 25 g
Catalog # COBBOS-7512-25G
Marketsource
Supplier: COMBI-BLOCKS
2-Hydroxy-3-nitropyridin
Catalog # COBBOS-7512-25G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 6332-56-5
Jump to:
Product Details & Documents
Synonyms: 3-Nitropyridin-2(1H)-one, 3-Nitro-2-pyridinol
- CAS Number:6332-56-5
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006270
- Molecular Weight:140.1 g/mol
- Melting Pt:224 °C
- EINECS:228-709-2
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:25 g



