4-Fluorindol
Glass bottle for solids, 25 g
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: 4-Fluoro-1H-indole
- CAS Number:387-43-9
- Formula:C₈H₆FN
- MDL Number:MFCD00055992
- Molecular Weight:135.14 g/mol
Documents
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:25 g



