ω-Chlor-2,4-difluoracetophenon
Glass bottle for solids, 25 g
Katalognr. COBBCA-4154-25G
Marketsource
Anbieter: COMBI-BLOCKS
ω-Chlor-2,4-difluoracetophenon
Katalognr. COBBCA-4154-25G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 51336-94-8
Direkt zu
Product Details & Documents
Synonyme: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone
- CAS Nummer:51336-94-8
- Formel:C₈H₅ClF₂O
- MDL-Nummer:MFCD00013252
- Molekulargewicht:190.58 g/mol
- Siedepunkt:233 °C (1013 hPa)
- Schmelzender Platingehalt:47…48 °C
- UN:3261
- ADR:8,II
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



