2-Hydroxyisonicotinic acid
Glass bottle for solids, 5 g
Catalog # COBBCA-4044-5G
Marketsource
Supplier: COMBI-BLOCKS
2-Hydroxyisonicotinic acid
Catalog # COBBCA-4044-5G
Supplier: COMBI-BLOCKS
Marketsource
CAS Number: 22282-72-0
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
- CAS Number:22282-72-0
- Formula:C₆H₅NO₃
- MDL Number:MFCD01318566
- Molecular Weight:139.11 g/mol
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



