2-Hydroxy-5-nitropyridin
Glass bottle for solids, 5 g
Katalognr. COBBPY-7173-5G
Marketsource
Anbieter: COMBI-BLOCKS
2-Hydroxy-5-nitropyridin
Katalognr. COBBPY-7173-5G
Anbieter: COMBI-BLOCKS
Marketsource
CAS Nummer: 5418-51-9
Direkt zu
Product Details & Documents
Synonyme: 5-Nitro-2-pyridinol
- CAS Nummer:5418-51-9
- Formel:C₅H₄N₂O₃
- MDL-Nummer:MFCD00006276
- Molekulargewicht:140.1 g/mol
- Siedepunkt:180…200 °C (0,2 mmHg)
- Schmelzender Platingehalt:191…192 °C
- EINECS:226-525-7
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



