Calorimetric standards
Benzoic acid, 2,0 gram pellets, bottle of 500, 500 g
Catalog # PARR3417
Supplier: PARR
Jump to:
Product Details & Documents
Calorific standard in powder or pellet form for standardising all oxygen bomb calorimeters.
Specifications
Specifications
- VE:500 g
- Bezeichnung:Benzoic acid, 2,0 gram pellets, bottle of 500