AuftragsverfolgungKontakt
Protein blocking buffers, Superior™ | Avantor
Protein blocking buffers, Superior™
Superior™ blocking buffer-dry blend in PBS, 5
Katalognr. 786-601
Anbieter:   G-Biosciences
Protein blocking buffers, Superior™
Katalognr. 786-601
Anbieter:  G-Biosciences
CAS Nummer:  
Protein blocking buffers, Superior™
Protein blocking buffers, Superior™
Superior™ blocking buffer-dry blend in PBS, 5
CAS Nummer:  
Preis
238.00 SFr.
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
Blocking buffer is an enhanced blocking agent available in multiple formats. Superior™ Blocking Buffer contains a proprietary antigenically non-determinant protein for blocking Western blot membranes, blocking and storage of ELISA plates, and blocking prior to immunohistochemistry staining.
Ideal for a high signal to background ratio in most systems and has a rapid blocking time of about two minutes for ELISA. It uses a non-serum protein and does not contain biotin or other animal source proteins that interfere with immuno-complexes. It is also suitable for assays that use avidin/streptavidin systems.
Technische Daten
Technische Daten
  • VE:
    5
  • Bezeichnung:
    Superior™ blocking buffer-dry blend in PBS