Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Gel drying solution for protein gels
Gel drying solution
  786-686
Gel drying solution for protein gels
  786-686
Gel drying solution for protein gels
Gel drying solution for protein gels
Gel drying solution
SFr. 394.00
1 * 1 gal US
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
  • Ready to use
  • Regulates rate of drying
  • Prevents cracking of the gel
  • Compatible with all drying membranes
  • Bezeichnung:
    Gel drying solution
  • Seq:
    03