Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Gel drying solution for protein gels | Avantor
Gel drying solution for protein gels
Gel drying solution
Katalognr. 786-685
Anbieter:   G-Biosciences
Gel drying solution for protein gels
Katalognr. 786-685
Anbieter:  G-Biosciences
Gel drying solution for protein gels
Gel drying solution for protein gels
Gel drying solution
Preis
SFr. 157.00
Einloggen für Verfügbarkeit
1 * 1 L
Product Details & Documents
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
  • Ready to use
  • Regulates rate of drying
  • Prevents cracking of the gel
  • Compatible with all drying membranes
Technische Daten
  • Bezeichnung:
    Gel drying solution
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.