Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Solid phase extraction cartridges, HyperSep™ Phenyl | Avantor

Solid phase extraction cartridges, HyperSep™ Phenyl

Kartusche, Phenyl, 40 - 60 µm, 3 ml, 200 mg
Katalognr. HYPE60108-387
Solid phase extraction cartridges, HyperSep™ Phenyl
Katalognr. HYPE60108-387
Solid phase extraction cartridges, HyperSep™ Phenyl
Solid phase extraction cartridges, HyperSep™ Phenyl

Kartusche, Phenyl, 40 - 60 µm, 3 ml, 200 mg

Preis
SFr. 375.00
Einloggen für Verfügbarkeit
1 * 50 ST
Product Details & Documents

Solid phase extraction cartridges, HyperSep™ phenyl offer alternative selectivity for retention of basic compounds. They are used for extraction of aromatic compounds and extraction of basic analytes.

  • Commonly employed for non-polar interactions
  • The electron density of aromatic ring provides different selectivity than other non-polar sorbents

HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.

Verpackung: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.

Technische Daten
  • Säulenform:
    Kartusche
  • Partikelgröße:
    40 - 60 µm
  • Kapazität:
    3 ml
  • Seq:
    02
  • Bettgewicht:
    200 mg
  • Phase:
    Phenyl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.