Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Affinity chromatography media, MabSelect Xtra™

MabSelect Xtra™, Liquid, 75 µm, 200 ml
  17-5269-02
 :   Cytiva
Affinity chromatography media, MabSelect Xtra™
  17-5269-02
 :  Cytiva
 :  
Affinity chromatography media, MabSelect Xtra™
Affinity chromatography media, MabSelect Xtra™

MabSelect Xtra™, Liquid, 75 µm, 200 ml

 :  
 
SFr. 7’820.00
   
1 * 200 mL
 

MabSelect Xtra has the same recombinant Protein A ligand as MabSelect but is based on a high-flow agarose base matrix with increased porosity and a slightly decreased particle size compared to MabSelect.

  • Higher dynamic binding capacity than currently available Protein A chromatography media
  • High purity capture due to high ligand selectivity and matrix hydrophilicity
  • Epoxy activation coupling method
  • Slightly lower pressure/flow properties compared to MabSelect SuRe and MabSelect
  • Improved process economics through reduced raw materials costs and/or reduced number of cycles
  • Prepacked, ready-to-use columns for convenient process development
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption

MabSelect Xtra is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect Xtra meets industrial demands with security of supply and comprehensive technical and regulatory support. The medium provides a lower overall production cost due to the possibility of processing concentrated feedstocks in fewer batches.

This results in higher capacity—MabSelect Xtra has about 30% higher dynamic binding capacity for IgG than currently available Protein A chromatography media. In addition, MabSelect Xtra has a higher capacitiy for many Fc-fusion proteins.

 

 

  • Partikelgröße:
    75 µm
  • VE:
    200 ml
  • Bezeichnung:
    MabSelect Xtra™
  • Typ:
    Liquid
  • Cat. No.:
    17-5269-02
  • Lagertemperatur:
    2 - 8 °C
  • pH-Bereich:
    3 - 10
  • Matrix:
    Highly cross-linked agarose
  • Ligand:
    Recombinant protein A (E.coli)
  • Fließrate:
    <300 cm/h
  • Bindungskapazität:
    40 mg human lgG/ml