AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™ | Avantor
Accessories for glove boxes, Precise™
IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
Katalognr. LBCP5242100
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Katalognr. LBCP5242100
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
Preis
SFr. 189.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Bezeichnung:
    IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
  • Seq:
    45