AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™ | Avantor
Accessories for glove boxes, Precise™
Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
Katalognr. LBCP5240100
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Katalognr. LBCP5240100
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
Preis
SFr. 536.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Technische Daten
  • Bezeichnung:
    Gas and vacuum 0,2 µm filter, traps particulates; for drying train or vacuum pump exhaust; 0,25" - 0,38" ID
  • Seq:
    26