AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
  135-1088
 :   LABCONCO
Accessories for glove boxes, Precise™
  135-1088
 :  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
SFr. 9’060.00
1 * 1 ST
  • Bezeichnung:
    Automatic pressure controller for 230 V units, incl. mounting bracket, hardware, 3/16" ID polyvinyl tubing, 3/32" ID polyvinyl tubing, and power cord, for automatic control of the main chamber pressure, main chamber purging and filling cycles, and transfer chamber evacuation and filling cycles
  • Seq:
    01