AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Katalognr. LBCP5245200
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Katalognr. LBCP5245200
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Preis
SFr. 315.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Bezeichnung:
    Gas and vacuum tubing connection kit, automatic pressure controller, with flexible vinyl tubing and fittings necessary to connect user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
  • Seq:
    03