Accessories for glove boxes, Precise™
Oxygen and moisture removal column, for reduceing the oxygen level to less than 5 ppm and moisture level to less than 50 ppm; requires connection to two manual valves on the main chamber and a diaphragm vacuum pump for atmosphere oxygen/moisture removal and recirculation
Catalog # LBCP5244200
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Bezeichnung:Oxygen and moisture removal column, for reduceing the oxygen level to less than 5 ppm and moisture level to less than 50 ppm; requires connection to two manual valves on the main chamber and a diaphragm vacuum pump for atmosphere oxygen/moisture removal and recirculation
- Seq:20