AuftragsverfolgungKontakt
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
Katalognr. LBCP5242501
Anbieter:   LABCONCO
Accessories for glove boxes, Precise™
Katalognr. LBCP5242501
Anbieter:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
Preis
696.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Bezeichnung:
    Oxygen and moisture removal tubing kit, incl. 25' fluorinated ethylene propylene (FEP) tubing, eight tube inserts and two connectors to connect the Oxygen and Moisture Removal Column to diaphragm pump
  • Seq:
    22