Accessories for glove boxes, Precise™
Flat work surface, black epoxy resin, may be placed atop two Protector Standard Base Cabinets to support Precise™ glove boxes, 1524×762×31,75 mm
Katalognr. LBCP4882807
Anbieter: LABCONCO
Direkt zu
Product Details & Documents
Technische Daten
- Bezeichnung:Flat work surface, black epoxy resin, may be placed atop two Protector Standard Base Cabinets to support Precise™ glove boxes, 1524×762×31,75 mm
- Seq:31