Control units
MIXcontrol 40, UK-Stecker
Catalog # 442-0773
Jump to:
Discontinued
Product Details & Documents
Specifications
- Bezeichnung:MIXcontrol 40, UK-Stecker
- Seq:10
- Cat. No.:442-0773
- Modell:MIXcontrol 40
- B×T×H:200×155×48 mm
- Drehzahlbereich:100 - 2000 min⁻¹
- Gewicht:1,9 kg
- Leistungsaufnahme:40 W