Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Diallylamin

10 kg
Catalog # 8.03521.9010
Flammable
Toxic
Corrosive
Supplier:   Merck
Diallylamin
Catalog # 8.03521.9010
Supplier:  Merck
CAS Number:  124-02-7
Diallylamin

10 kg

CAS Number:  124-02-7
Price
Inquire For Price
Log in for availability
1 * 10 kg
Product Details & Documents
  • CAS Number:124-02-7
  • Formula:C₆H₁₁N
  • MDL Number:MFCD00008642
  • Molecular Weight:97.16 g/mol
  • Storage Temperature:Raumtemperatur
  • ADR:3 (6.1, 8),II
  •  :M-Clarity™ Qualitätsstufe = MQ200
  • EINECS:204-671-2
  • UN:2359
Specifications

Specification Test Results

  • Solubility:
    86 g/l (20 °C)
  • Melting Point:
    -88 °C
  • Molar Mass:
    97.15 g/mol
  • Boiling Point:
    111 °C (1013 hPa)
  • Vapor Pressure:
    24.2 hPa (20 °C)
  • Flash Point:
    11 °C
  • Refractive Index:
    1.4405 (20 °C, 589 nm)
  • Explosion Limit:
    0.93 - 14.2 %(V)
  • Density:
    0.79 g/cm3 (20 °C)
  • pH:
    11.5 (9.7 g/l, H2O)
  • Ignition Point:
    273 °C

Specifications

  • VE:
    10 kg

Specifications