AuftragsverfolgungKontakt
Spring scissors, 6 mm cutting edge
Sharp, straight tips, 140 mm, 6 mm, 0,225 mm
  FSCI15020-15
Marketsource
Spring scissors, 6 mm cutting edge
  FSCI15020-15
Marketsource
Spring scissors, 6 mm cutting edge
Spring scissors, 6 mm cutting edge
Sharp, straight tips, 140 mm, 6 mm, 0,225 mm
SFr. 229.03
1 * 1 ST
These scissors are made of stainless steel.
  • Typ:
    Sharp, straight tips
  • Länge:
    140 mm
  • Seq:
    02
  • Spitzengröße:
    0,225 mm
  • Spitzenlänge:
    6 mm