Abdeckmatten für Deep-Well-Platten
Verschlussmatten, TPE, für 96-Well-Platten (1,2 ml), Low Profile
Katalognr. 738-1002
Anbieter: Brand
Direkt zu
Product Details & Documents
Cover mats reduce the maximum volume of wells.
Dokumente
Brand Deep-well Plates
Produktliteratur
Technical Note BRAND PCR sealing options
Anwendungshinweise
Technische Daten
- Bezeichnung:Verschlussmatten, TPE, für 96-Well-Platten (1,2 ml), Low Profile
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.