Degustation and drinking cups
70 ml, 65 mm
Katalognr. SEMA5651
Anbieter: SEMADENI
Direkt zu
Product Details & Documents
Polystyrene, clear.
- Good chemical resistance
- Good temperature resistance
These dugustation and drinking cups are crystal clear, sprayed design, well standing, stable and elegant shape with content marking.
Technische Daten
- Kapazität:70 ml
- Höhe:65 mm
- Seq:01



