Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Degustation and drinking cups | Avantor

Degustation and drinking cups

200 ml, 96 mm
Katalognr. SEMA5653
Anbieter:   SEMADENI
Degustation and drinking cups
Katalognr. SEMA5653
Anbieter:  SEMADENI
Degustation and drinking cups

200 ml, 96 mm

Preis
SFr. 136.00
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents

Polystyrene, clear.

  • Good chemical resistance
  • Good temperature resistance

These dugustation and drinking cups are crystal clear, sprayed design, well standing, stable and elegant shape with content marking.

Technische Daten
  • Kapazität:
    200 ml
  • Höhe:
    96 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.