Drenchers
Drencher, bent, 203 mm, 3,0 mm
SOCO376.30203B
These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.
- Luer lock fittings
- Ext.Ø:3,0 mm
- Bezeichnung:Drencher, bent
- Länge:203 mm
- Seq:04



