Cover glasses
#1,5, 18 mm
Katalognr. HECH92101401120
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
High spectral transmission, excellent flatness, optimum resistance to chemicals.
- Free from colour distortion borosilicate glass D 263™ M
- Excellent quality: Low levels of inclusions, striae, bubbles and streaks
Zertifizierung: ISO 8255
Verpackung: In tight-closing plastic boxes for protection against damp and climatic influences.
Technische Daten
- Ø:18 mm
- Nr.:#1,5
- Seq:01