Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 6400 series, width 110 mm, BBP™11, 70 m, Schwarz
Catalog # BRDY804778
Supplier: Brady
Jump to:
Product Details & Documents
Specifications
- Farbe:Schwarz
- Bezeichnung:Thermal transfer printer ribbon, 6400 series, width 110 mm
- Länge:70 m
- Seq:17
- Für:BBP™11