Thermal transfer printer labels, type B-449
Removable printer labels, B-449, THT-19-449-1, PP, Acryl, Thermal transfer, Weiß, 360X-Plus II, 76×50 mm, 1000 labels/roll
BRDY067113
: Brady
Printable, removable PP labels for thermal transfer printers.
- Druckermodell:360X-Plus II
- Printing method:Thermal transfer
- Typ:B-449
- Farbe:Weiß
- Bezeichnung:Removable printer labels
- Abmessungen:76×50 mm
- Material:PP
- Seq:06
- Modell:THT-19-449-1
- Verpackt:1000 labels/roll
- Klebstoff:Acryl



