Thermal transfer printer labels, type B-8423
Printer labels, B-8423, BPT-7-8423-1, Polyester, Permanent, Acryl, Thermal transfer, Weiß, BBP12, 69×31 mm, 1000 labels/roll
Katalognr. BRDY236717
Anbieter: Brady
Direkt zu
Product Details & Documents
Printable satin polyester labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Technische Daten
- Druckermodell:BBP12
- Printing method:Thermal transfer
- Typ:B-8423
- Farbe:Weiß
- Bezeichnung:Printer labels
- Abmessungen:69×31 mm
- Material:Polyester
- Seq:10
- Modell:BPT-7-8423-1
- Verpackt:1000 labels/roll
- Klebstoff:Permanent, Acryl
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



