Jars, glass
Glass jar with metal locking bracket and rubber seal around the rim, 500 ml
Katalognr. SPVAH03064
Anbieter: GRAVIS
Direkt zu
Product Details & Documents
Soda lime glass jars, suitable for storage of samples in the laboratory.
Technische Daten
- Kapazität:500 ml
- Bezeichnung:Glass jar with metal locking bracket and rubber seal around the rim
- Seq:01