Iodmonochlorid
Glass bottle for solids, 25 g
CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 Physical Form: Crystalline/Liquid Density: 3.1822 Sensitivity: Moisture and Light Sensitive Appearance: Dark red/black Odour: Pungent Melting Point: 25-27° Boiling Point: 96-98° Solubility: Soluble in water, alcohol, ether, carbon disulfide Beilstein Reference: 3902972 Merck Reference: 14,5017 UN No.: 1792 MDL No.: MFCD00011354
: Iodochloride
- :7790-99-0
- :ClI
- :MFCD00011354
- :162.36 g/mol
- :14,5017
- :96…98 °C
- :25…27 °C
- :3,1822
- :Raumtemperatur
- :232-236-7
- :1792
- :8,II
- Packung:Glass bottle for solids
- VE:25 g