AuftragsverfolgungKontakt
Iodmonochlorid
Glass bottle for solids, 25 g
  39104.14
Iodmonochlorid
  39104.14
 :  7790-99-0
Iodmonochlorid
Iodmonochlorid
Glass bottle for solids, 25 g
 :  7790-99-0
55.80 SFr.
1 * 25 g
CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 Physical Form: Crystalline/Liquid Density: 3.1822 Sensitivity: Moisture and Light Sensitive Appearance: Dark red/black Odour: Pungent Melting Point: 25-27° Boiling Point: 96-98° Solubility: Soluble in water, alcohol, ether, carbon disulfide Beilstein Reference: 3902972 Merck Reference: 14,5017 UN No.: 1792 MDL No.: MFCD00011354
 :  Iodochloride
  •  :7790-99-0
  •  :ClI
  •  :MFCD00011354
  •  :162.36 g/mol
  •  :14,5017
  •  :96…98 °C
  •  :25…27 °C
  •  :3,1822
  •  :Raumtemperatur
  •  :232-236-7
  •  :1792
  •  :8,II
  • Packung:
    Glass bottle for solids
  • VE:
    25 g