AuftragsverfolgungKontakt
Pre-prepared polystyrene kits, EasiCal™ | Avantor
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-1, 580 - 7500000 g/mol, 5
Katalognr. VARIPL2010-0505
Anbieter:   VARIAN
Pre-prepared polystyrene kits, EasiCal™
Katalognr. VARIPL2010-0505
Anbieter:  VARIAN
CAS Nummer:  
Pre-prepared polystyrene kits, EasiCal™
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-1, 580 - 7500000 g/mol, 5
CAS Nummer:  
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
  • Easy three step process with no fuss
  • Cost effective
  • Only two injections for improved productivity
  • Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.
Technische Daten
Technische Daten
  • VE:
    5
  • Bezeichnung:
    Polystyrene PS-1, 580 - 7500000 g/mol