Wrenches
Wrench for jerrycan closures, DIN 51, 5 l / 10 l, PE
Katalognr. DECO60025
Anbieter: Borer Chemie
Direkt zu
Product Details & Documents
These wrenches are used for jerrycan closures.
Technische Daten
- Bezeichnung:Wrench for jerrycan closures, DIN 51, 5 l / 10 l
- Material:PE
- Seq:02