Glass Microfiber Filter Paper
GF 6, 90 mm
PRATA090875
These glass microfiber filter papers allow fast filtration and high particle retention. They can be used at temperatures up to 380 °C.
- Chemical compatibility with many solvents and with reagents
- Suitable for microfiltration
- Sorte:GF 6
- Seq:3
- Größe:90 mm
- Cat. No.:PRATA090875
- Stärke:480 µm
- Gewicht:75 g/m²



