Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
HPLC columns, HyperREZ™ XP Carbohydrate Pb2+ | Avantor

HPLC columns, HyperREZ™ XP Carbohydrate Pb2+

Analytisch, 8 µm, 7,70 mm, 50 mm
Katalognr. HYPE69008-057726
HPLC columns, HyperREZ™ XP Carbohydrate Pb2+
Katalognr. HYPE69008-057726
HPLC columns, HyperREZ™ XP Carbohydrate Pb2+

Analytisch, 8 µm, 7,70 mm, 50 mm

Preis
SFr. 1’300.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These polymer-based columns for carbohydrate analysis have benefits in temperature and pH stability and enhanced solvent choices.

  • Designed for the determination of carbohydrates, saccharides, organic acids and alcohols
  • Efficient and reproducible monodisperse particles
  • Stable for long column lifetimes even at low pH and high temperatures
Technische Daten
  • Säulenform:
    Analytisch
  • Ø int.:
    7,70 mm
  • Partikelgröße:
    8 µm
  • Länge:
    50 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.