Microsampling tools
Straight micro probe
Catalog # PERCN9302605
Supplier: PerkinElmer
Jump to:
Product Details & Documents
This microsampling tool kit is suitable for FTIR microscopy.
Delivery information: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.
Specifications
- Bezeichnung:Straight micro probe
- Seq:04