Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Drenchers | Avantor

Drenchers

Drencher, straight, 51 mm, 1,2 mm
Katalognr. SOCO376.1251S
Anbieter:   Socorex Isba
Drenchers
Katalognr. SOCO376.1251S
Anbieter:  Socorex Isba
Drenchers
Drenchers

Drencher, straight, 51 mm, 1,2 mm

Preis
SFr. 52.70
Einloggen für Verfügbarkeit
1 * 3 ST
Product Details & Documents

These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.

  • Luer lock fittings
Technische Daten
  • Ext.Ø:
    1,2 mm
  • Bezeichnung:
    Drencher, straight
  • Länge:
    51 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.