L-Methionin
Glasflasche für Feststoffe, 100 g
Anbieter: PanReac AppliChem
Direkt zu
Product Details & Documents
Synonyme: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- CAS Nummer:63-68-3
- Formel:C₅H₁₁NO₂S
- MDL-Nummer:MFCD00063097
- Molekulargewicht:149.21 g/mol
- Merck-Index:13,06004
- Siedepunkt:278 °C (1013 hPa)
- Schmelzender Platingehalt:136 °C
- Dichte:1,373 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-562-9
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



