Buffer solutions, according to DIN 19266, Fluka™
9,180, —, Borat, 25 °C, Plastic bottle, 6×1 L, 6 L
Katalognr. 33595-6X1L
Anbieter: Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Katalognr. 33595-6X1L
Anbieter: Honeywell Chemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Buffers with fungicide (except pH 10,012)
Certifications: According to DIN 19266.
Documents
Technische Daten
Technische Daten
- Packung:Plastic bottle
- pH-Puffertyp:Borat
- VE:6 L
- Referenztemperatur:25 °C
- pH Genauigkeit:—
- Pk Info:6×1 L
- pH-Wert:9,180
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



