Bajonettfilter, X-plore®
Filterpad-Kappe
Katalognr. 111-1046
Anbieter: Dräger
Direkt zu
Product Details & Documents
Sortiment an Filtern mit einem Dräger-spezifischen Bajonettanschluss für X-plore® 3300/3500 Halbmasken und X-plore® 5500 Vollmasken.
- High level of user safety, great fit under face shields and visors
- Robust gas filter with a blue ABS plastic housing (boiling point >65 °C)
- White particle filter housing for easy identification of P3 filters
- Option to upgrade a gas filter to a combination filter by simply adding a P1 or P2 pad or Dräger X-plore® Pure filter
- Safe, quick and easy bayonet connection with clear guidance for positioning the filter securely
- Filters inserted top-down for secure positioning
- Shelf life (from date of production): Gas and combination filters 6 years, particle filters 12 years, pads 3 years
- Prefilters, to protect combination filters against clogging and blocking
Zertifizierung: CE-zertifiziert (EN 14387, EN 143:2000/A1:2006); Beschriftung „R“ (Kombifilter): zertifiziert für die Wiederverwendung (mehr als eine Schicht)
Zertifiziert nach australischem Standard AS/NZS 1716 (SAI Global)
Technische Daten
- Typ:Filterpad-Kappe
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.