Packaging Bags
0,09 mm, 150×200 mm
Katalognr. SCAC3002
Anbieter: SCANCELL FOLIEN VETRIEB
Direkt zu
Product Details & Documents
These vacuum bags are with sealed edge and ideal for packaging of gas-sensitive goods and foods.
- PA/PE, transparent
Technische Daten
- Abmessungen:150×200 mm
- Seq:02
- Stärke:0,09 mm